• USA Home
  • M3277 - Methoxypolyethylene glycol activated with cyanuric chloride

EMAIL THIS PAGE TO A FRIEND
M3277 Sigma

Methoxypolyethylene glycol activated with cyanuric chloride

average mol wt 5,000

  • Linear Formula CH3O(CH2CH2O)n-C3N3Cl2

  •  MDL number MFCD00131596

Purchase

Description

Packaging

1, 5 g in poly bottle

Application

Methoxypolyethylene glycol activated with cyanuric chloride has been used in a study to assess the biophysical consequences of linker chemistry and polymer size on stealth erythrocytes. It has also been used in a study to quantify dithiothreitol displacement of functional ligands from gold nanoparticles. This detergent is also for modification of proteins; it may improve solubility and extend biological lifetime.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?