EMAIL THIS PAGE TO A FRIEND
M3383 Sigma

Metrizamide

Grade I, lyophilized powder

Synonym: 2-[3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzamido]-2-deoxy-D-glucose, Metrizamide, mixture of anomers

Purchase

Properties

type   Grade I
assay   ~99%
form   lyophilized powder
mp   222-224 °C (dec.)(lit.)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
LZ6650000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

D1556 OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
D2158 Histodenz, nonionic density gradient medium

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?