• USA Home
  • M3386 - 1-Methylhistamine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
M3386 Sigma

1-Methylhistamine dihydrochloride

Synonym: 1-Methyl-4-(β-aminoethyl)imidazole dihydrochloride

  • CAS Number 6481-48-7

  • Empirical Formula (Hill Notation) C6H11N3 · 2HCl

  • Molecular Weight 198.09

  •  MDL number MFCD00037000

  •  PubChem Substance ID 329817972

Purchase

Properties

InChI Key   AGXVEALMQHTMSW-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Biochem/physiol Actions

Major metabolite of histamine by histamine N-methyltransferase.

Linkage

Similar to M 4910, but prepared for Sigma.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M4910 1-Methylhistamine dihydrochloride, ≥98% (TLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?