• USA Home
  • M3398 - α2-Macroglobulin from human blood plasma

EMAIL THIS PAGE TO A FRIEND
M3398 Sigma

α2-Macroglobulin from human blood plasma

Prestained

Purchase

Properties

Related Categories Biochemicals and Reagents, Coagulation Proteins and Reagents, Plasma & Blood Proteins, Proteins and Derivatives
mol wt   unstained protein mol wt 180,000 Da
storage temp.   2-8°C
Gene Information   human ... A2M(2)

Description

Biochem/physiol Actions

α2-Macroglobulin (α2M) is a multifunctional protein that is a broad spectrum protease inhibitor. α2M is a large homotetrameric glycoprotein (720 kDa) that is connected by disulfide-linked dimers which non-covalently interact to give the quaternary structure.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?