• USA Home
  • M3411 - Marker for 2D Electrophoresis

EMAIL THIS PAGE TO A FRIEND
M3411 Sigma

Marker for 2D Electrophoresis

urea solution

Purchase

Properties

Related Categories 2D Markers, Core Bioreagents, Isoelectric Focusing and Two Dimensional Electrophoresis, Life Science Reagents for Protein Electrophoresis, Markers,
form   urea solution
usage   1 vial sufficient for 20-40 applications (on gels to be stained with Brilliant Blue)
mol wt   mol wt 17,000-89,000 Da
shipped in   dry ice
storage temp.   −70°C

Description

Application

This marker is specifically designed for two-dimensional electrophoresis (2D) and denaturing (urea) IEF. The four proteins in this marker have been selected to provide a diagonal pattern across a 2D gel. The pI range is 7.6 to 3.8.

Packaging

Vial contains approx. 200 μl of a mixture of four proteins.

Physical form

Solution in 8 M urea, 2% 2-mercaptoethanol

Price and Availability


Amicon Pro Purification System
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?