• USA Home
  • M3552 - Macrophage Inflammatory Protein-3β human

EMAIL THIS PAGE TO A FRIEND
M3552 Sigma

Macrophage Inflammatory Protein-3β human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Synonym: ELC, MIP-3β

Purchase

Properties

Related Categories Cell Biology, Cell Culture, Cell Signaling and Neuroscience, Chemokines, Cytokines and Growth Factors,
recombinant   expressed in E. coli
assay   ≥97% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
suitability   cell culture tested
storage temp.   −20°C
Gene Information   human ... CCL19(6363)

Description

Analysis Note

The biological activity is measured by using BAF3 cells transfected with human CCR7.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoracetic acid containing 1.25 mg of bovine serum albumin.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?