• USA Home
  • M3638 - 1-Methylcyclopropanecarboxylic acid ethyl ester

EMAIL THIS PAGE TO A FRIEND
M3638 Sigma

1-Methylcyclopropanecarboxylic acid ethyl ester

~98%

Synonym: Ethyl 1-methylcyclopropanecarboxylate

Purchase

Properties

assay   ~98%
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
87.8 °F
Flash Point(C) 
31 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?