• USA Home
  • M3639 - Maltooligosaccharide from corn syrup

EMAIL THIS PAGE TO A FRIEND
M3639 Sigma

Maltooligosaccharide from corn syrup

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

Maltose linear homologs G4 through G10.
Essentially free of G1 through G3 (glucose, maltose and maltotriose).

Substrates

Reported to be useful as a substrate for α-amylase.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

M8128 Maltopentaose, ≥95%
M7753 Maltoheptaose, ≥60% (HPLC)
M9153 Maltohexaose, ≥65% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?