• USA Home
  • M3653 - 5-Methoxytryptamine-d4 hydrochloride

EMAIL THIS PAGE TO A FRIEND
M3653 Sigma

5-Methoxytryptamine-d4 hydrochloride

Synonym: 3-(2-Amino[ethyl-::FId::FN4])-5-methoxyindole, O-Methylserotonin-::FId::FN4

Purchase

Properties

isotopic purity   98 atom % D
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?