• USA Home
  • M3898 - Methyl 2-O-α-D-mannopyranosyl-α-D-mannopyranoside

EMAIL THIS PAGE TO A FRIEND
M3898 Sigma

Methyl 2-O-α-D-mannopyranosyl-α-D-mannopyranoside

~95%

Synonym: α-D-Man-[1→2]-α-D-Man-1→OMe

Purchase

Properties

InChI Key   YUAFWDJFJCEDHL-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Description

Application

Model compound for NMR studies of manno-oligosaccharides.

Substrates

Substrate for the study of linkage-specific 1,2-α-mannosidase from Aspergillus niger.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?