• USA Home
  • M4046 - Mitogen-Activated Protein Kinase 2 fragment 326-339

EMAIL THIS PAGE TO A FRIEND
M4046 Sigma

Mitogen-Activated Protein Kinase 2 fragment 326-339

≥97% (HPLC)

Synonym: P38RK Substrate

  • CAS Number 171869-94-6

  • Empirical Formula (Hill Notation) C72H126N22O19

  • Molecular Weight 1603.91

  •  MDL number MFCD00677328

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Lys-Val-Pro-Gln-Thr-Pro-Leu-His-Thr-Ser-Arg-Val-Leu-Lys

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?