• USA Home
  • M4058 - SigmaScreen Streptavidin coated plates

EMAIL THIS PAGE TO A FRIEND
M4058 Sigma

SigmaScreen Streptavidin coated plates

96-well, solid black

Purchase

Properties

Related Categories Cell Culture, Coated Plates, Drug Discovery More...
material   clear polystyrene
description   flat-bottom
storage temp.   2-8°C

Description

General description

SigmaScreen Streptavidin coated plates provide superior binding capacity with the greatest signal to noise ratios for improved sensitivity. With at least 6 pmol/well for the 96 well plates and approximately 4 pmol/well for the 384 well plates, SigmaScreen Streptavidin coated plates are pre-blocked with BSA and developed for maximum binding of biotinylated nucleic acids, antibodies, proteins and peptides. Manufactured under ISO9002 in Sigma′s GMP facility, Sigma′s Streptavidin coated plates deliver high well-to-well and plate-to-plate consistency.

Legal Information

SigmaScreen is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?