• USA Home
  • M4545 - L-Methionine γ-lyase from Pseudomonas putida

EMAIL THIS PAGE TO A FRIEND
M4545 Sigma

L-Methionine γ-lyase from Pseudomonas putida

lyophilized powder, pkg of ≥1.0 units/vial

Purchase

Properties

Related Categories 4.4.x.x C-S bonds, 4.x.x.x Lyases, Amino Acid Metabolism, Biochemicals and Reagents, Cell Biology,
form   lyophilized powder
packaging   pkg of ≥1.0 units/vial
storage temp.   −20°C

Description

Unit Definition

One unit will release 1.0 μmol of α-ketobutyrate from L-methionine per min at pH 8.0 at 37 °C.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?