• USA Home
  • M4757 - Microperoxidase (MP-8) sodium salt from equine heart cytochrome c

EMAIL THIS PAGE TO A FRIEND
M4757 Sigma

Microperoxidase (MP-8) sodium salt from equine heart cytochrome c

~90% (HPLC)

  • CAS Number 104780-81-6

  • Empirical Formula (Hill Notation) C68H87FeN15O17S2

  • Molecular Weight 1506.48

  •  MDL number MFCD00131609

Purchase

Description

Application

Of possible interest in cell permeability studies and in electron microscopic immunocytochemistry.

Biochem/physiol Actions

Has peroxidase activity.

Other Notes

The proposed structure of microperoxidase is the heme portion of cytochrome c with amino acids 14 through 21 still attached.

Preparation Note

Prepared by enzymatic degradation.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?