• USA Home
  • M4809 - Matrix Metalloproteinase 9 human

EMAIL THIS PAGE TO A FRIEND
M4809 Sigma

Matrix Metalloproteinase 9 human

>85% (SDS-PAGE), recombinant, expressed in transfected cells, buffered aqueous solution

Synonym: 95 kDa Gelatinase, Gelatinase-B, MMP-9

Purchase

Description

Biochem/physiol Actions

Degrades the collagens which make up the extracellular matrix which has a role in the control of angiogenesis.

Physical form

Solution in 5 mM Tris-HCl, pH 7.5, and 0.1 mM calcium chloride containing 0.005% Brij 35.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M8945 Matrix Metalloproteinase-9 human, recombinant, expressed in NSO cells, >95% (SDS-PAGE), buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?