EMAIL THIS PAGE TO A FRIEND
M4815 Sigma

MMP Control-4

buffered aqueous solution

Purchase

Description

Analysis Note

Recommended use for immunoblotting: mix 1:1 with 2X SDS-PAGE buffer and apply 20-40 μl per lane.

Application

Used as a qualitative positive control in immunoblotting for MMP-12.

General description

Concentrated, serum-free, cell culture media from human fibroblasts containing human macrophage elastase (MMP-12), as well as other MMPs and TIMPs.

Physical form

Solution in 0.01 M phosphate buffered saline

Price and Availability


Better Together

Discover your Target

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?