• USA Home
  • M5009 - N-Msoc-L-proline (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
M5009 Sigma

N-Msoc-L-proline (dicyclohexylammonium) salt

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?