• USA Home
  • M5010 - L-Methionine−Αgarose

EMAIL THIS PAGE TO A FRIEND
M5010 Sigma

L-Methionine−Αgarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Amino Acid Resins, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
extent of labeling   2-10 μmol per mL
matrix   cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino
matrix spacer   1 atom
storage temp.   2-8°C

Description

Physical form

Suspension in 2.0 M NaCl

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?