• USA Home
  • M5307 - SigmaScreen Poly-D-lysine coated plates

EMAIL THIS PAGE TO A FRIEND
M5307 Sigma

SigmaScreen Poly-D-lysine coated plates

96-well, black/clear bottom

Purchase

Properties

Related Categories Cell Culture, Cell Culture Supplies, Cell Culture Surfaces, Coated Plates, Drug Discovery,
description   flat bottom
feature   lid
storage temp.   2-8°C

Description

General description

SigmaScreen Poly D-Lysine (PDL) coated multiwell plates are coated with a synthetic PDL molecular weight range of 70-150 kDa. By coating the negatively charged polystyrene surface with PDL, the resulting positively charged surface has been shown to improve cell survival under low serum or serum-free conditions. SigmaScreen PDL coated multiwell plates can be used with a variety of cell lines including HEK-293 and mouse 3T3. For a more comprehensive listing of cell lines used with PDL coated multiwell plates, please refer to the PDL coated plate technical bulletin. Manufactured under ISO9002 in Sigma′s GMP facility, Sigma′s PDL coated multiwell plates are bioburden tested (fungal and bacterial) and endotoxin free.

Legal Information

SigmaScreen is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?