• USA Home
  • M5311 - Greiner high and medium binding 96 well plates

EMAIL THIS PAGE TO A FRIEND
M5311 Sigma

Greiner high and medium binding 96 well plates

clear conical bottom

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Purchase

Properties

Related Categories 96 Well Plates, Cell Culture, Drug Discovery, Greiner 96-Well Multiwell Plates, Labware,
material   clear conical bottom
  polystyrene
sterility   non-sterile
feature   lid: no
packaging   case of 40
mfr. no.   Greiner 655085
size   96 wells
color   clear plate
matrix active group   high binding surface treatment

Description

General description

MICROLON® 600 96W Microplate.

Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Other Notes

96 well C- bottom; high binding microplate, without lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Suitability

UsefμL in immunological assays. MICROLON 600 is a hydrophobic medium binding surface that will bind up to 600 ng/cm2 of non-polar proteins and peptides. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?