• USA Home
  • M5392 - Metallothionein II from rabbit liver

EMAIL THIS PAGE TO A FRIEND
M5392 Sigma

Metallothionein II from rabbit liver

lyophilized powder

Purchase

Properties

form   lyophilized powder
Metal as Cd + Zn content   ~7%
impurities   salt, essentially free
storage temp.   2-8°C

Description

Linkage

This is the form that elutes last from an anion exchange column and is probably identical to Form I of Nordberg, et al.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?