EMAIL THIS PAGE TO A FRIEND
M5406 Sigma

Methaqualone-d4

Synonym: 2-Methyl-3-(2-methylphenyl)-4(3H)-quinazolinone-5,6,7,8-d4

  • CAS Number 60124-85-8

  • Empirical Formula (Hill Notation) C16H10D4N2O

  • Molecular Weight 254.32

  •  MDL number MFCD00079423

Purchase

Properties

InChI Key   JEYCTXHKTXCGPB-DOGSKSIHSA-N
form   neat
drug control   Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
mass shift   M+4
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?