• USA Home
  • M5432 - SigmaScreen Streptavidin coated plates
EMAIL THIS PAGE TO A FRIEND
M5432 Sigma
SigmaScreen Streptavidin coated plates
96-well, clear
Purchase
Description
General description
SigmaScreen Streptavidin coated plates provide superior binding capacity with the greatest signal to noise ratios for improved sensitivity. With at least 6 pmol/well for the 96 well plates and approximately 4 pmol/well for the 384 well plates, SigmaScreen Streptavidin coated plates are pre-blocked with BSA and developed for maximum binding of biotinylated nucleic acids, antibodies, proteins and peptides. Manufactured under ISO9002 in Sigma′s GMP facility, Sigma′s Streptavidin coated plates deliver high well-to-well and plate-to-plate consistency.
Legal Information
SigmaScreen is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Detect Your Unseen Protein
All labs need water
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?