• USA Home
  • M5436 - Greiner high and medium binding 96 well plates

EMAIL THIS PAGE TO A FRIEND
M5436 Sigma

Greiner high and medium binding 96 well plates

clear polystyrene wells flat bottom (with micro-clear bottom), surface treatment high binding, non-sterile

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Purchase

Properties

Related Categories 96 Well Plates, Antibody Plates, Antibody/Immunology Supplies, Greiner, Greiner 96-Well Multiwell Plates,
material   clear polystyrene wells flat bottom (with micro-clear bottom)
  white polystyrene plate
sterility   non-sterile
packaging   case of 40
mfr. no.   Greiner 655094
maximum volume   392 μL
size   96 wells
working volume   340 μL
matrix active group   high binding surface treatment

Description

General description

LUMITRAC 600 96W Microplate, well working volume 25-340 μL; total well volume 392 μL.

Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Other Notes

96 well flat (F) μClear® bottom; high binding microplate, chimney style, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Suitability

White plates minimize cross talk in luminescence procedures. LUMITRAC 600 will bind up to 600 ng/cm2. Clear bottoms allow for monitoring thru the plate bottom. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

μClear is a registered trademark of Greiner Bio-One GmbH

LUMITRAC is a trademark of Greiner Bio-One GmbH

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?