• USA Home
  • M5520 - Myokinase from chicken muscle

EMAIL THIS PAGE TO A FRIEND
M5520 Sigma

Myokinase from chicken muscle

essentially salt-free, lyophilized powder, 1,500-3,000 units/mg protein (biuret)

Synonym: ATP: AMP phosphotransferase, Adenylate Kinase

Purchase

Properties

Related Categories 2.7.x.x Phosphorus containing groups, 2.x.x.x Transferases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   essentially salt-free, lyophilized powder
foreign activity   3-phosphoglyceric phosphokinase ≤0.1%
  lactic dehydrogenase ≤0.02%
  pyruvate kinase ≤0.05%
storage temp.   −20°C

Description

Unit Definition

One unit will convert 2.0 μmoles of ADP to ATP+AMP per min at pH 7.6 at 37°C.

General description

Mutational analysis of the amino acid proline 17 of myokinase from chicken muscle is critical for structural stability, substrate binding and enzyme activity.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Protocols

Enzymatic Assay of Myokinase (EC 2.7.4.3)

Replaces OP SP-ADP01. New document assigned to conform to current document numbering system. Refer to CR SOP-DEK-ENZ47.
Keywords: Extinction coefficient, Molecular biology

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?