• USA Home
  • M5529 - Morantel (+)-tartrate salt

EMAIL THIS PAGE TO A FRIEND
M5529 Sigma

Morantel (+)-tartrate salt

Synonym: 1,4,5,6-Tetrahydro-1-methyl-2-(2-[3-methyl-2-thienyl]ethenyl)pyrimidine

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics G-M, Antiparasitic / Antiprotozoal / Anthelminthic, Biochemicals and Reagents,
InChI Key   GGXQONWGCAQGNA-UUSVNAAPSA-N
color   light yellow
solubility   H2O: soluble4.0 mL, hazy, light yellow (200 mg + 4.0 mL H2O)
Mode of action   enzyme | inhibits

Description

Application

Morantel (+)-tartrate is used to study allosteric modulation in neuronal nicotinic acetylcholine receptors. It is also used as a bovine dewormer.

Biochem/physiol Actions

Morantel is a potent non-canonical (binding site) acetylcholine (ACh) receptor agonist and inhibits fumarate reductase.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UW0246000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?