• USA Home
  • M5531 - 19-Methylarachidic acid

EMAIL THIS PAGE TO A FRIEND
M5531 Sigma

19-Methylarachidic acid

~98% (capillary GC)

Synonym: 19-Methyleicosanoic acid

Purchase

Properties

Related Categories Biochemicals and Reagents, Branched Fatty Acids, Fatty Acids and conjugates, Fatty Acyls, Lipids More...
InChI Key   BDGYZTCVQAZQFG-UHFFFAOYSA-N
assay   ~98% (capillary GC)
storage temp.   2-8°C

Description

Packaging

10, 25, 100 mg in ampule

Biochem/physiol Actions

19-Methyleicosanoic acid (19-MEA) is related to 18-methyleicosanoic acid (18-MEA), a fatty acid found covalently associated with fur and hair. 19-Methyleicosanoic acid may be used in comparative studies with 18-MEA and other methyleicosanoic acids that affect hair and fur fiber structure. 19-MEA may be studied for use as a component of hair treatment medications and formulations.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?