• USA Home
  • M5628 - 6-Mercaptopurine-2′-deoxyriboside

EMAIL THIS PAGE TO A FRIEND
M5628 Sigma

6-Mercaptopurine-2′-deoxyriboside

Synonym: 2′-Deoxy-6-thioinosine, dI6S

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   WACLJMMBCDNRJE-UHFFFAOYSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

6-Mercaptopurine-2′-deoxyriboside (2′-deoxy-6-thioinosine; dI6S) is a thiolated inosine deoxynucleoside that may possess antitumor activity.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UP0762000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?