• USA Home
  • M5662 - Monocyte chemotactic and activating factor, Human

EMAIL THIS PAGE TO A FRIEND
M5662 Sigma

Monocyte chemotactic and activating factor, Human

recombinant, expressed in E. coli

Synonym: MCAF

Purchase

Properties

recombinant   expressed in E. coli
shipped in   dry ice
storage temp.   −20°C

Description

Quantity

Maximal chemotactic activity is achieved at 10-100 ng/mL medium.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M6667 Monocyte Chemotactic Protein-1 human, ≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?