• USA Home
  • M5897 - Myosin subfragment 2 from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
M5897 Sigma

Myosin subfragment 2 from rabbit muscle

buffered aqueous glycerol solution

Purchase

Properties

Related Categories Biochemicals and Reagents, Myosin, Proteins and Derivatives, Structural Proteins
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C

Description

Preparation Note

"Long Subfragment-2," produced from a chymotryptic digest of heavy meromyosin.

Quality

Significantly free of ATPase activity.

Physical form

Solution in 50% glycerol containing 0.5 M potassium chloride and 0.025 M potassium phosphate, pH 6.2

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?