• USA Home
  • M5928 - Anti-Matrix Metalloproteinase-2, Propeptide Region antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
M5928 Sigma

Anti-Matrix Metalloproteinase-2, Propeptide Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-MMP-2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Adhesion, Attachment, and Matrix Factors, Antibodies against Adhesion/Attachment/Matrix Factors, Antibodies for Cell Biology,
species reactivity   human
application(s)   immunohistochemistry (frozen sections): suitable
  immunoprecipitation: suitable
  indirect ELISA: suitable
  western blot: 1:1,000
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
mol wt   antigen mol wt 79 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... MMP2(4313)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the propeptide region of human matrix metalloproteinase-2 (gelantinase-A).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 15 mM sodium azide and 50% glycerol.

Specificity

Reacts with reduced and native MMP-2 free or bound to TIMP-2. Recognizes only the latent form of MMP-2 and not the active forms. May be used to distinguish the zymogen and the activated enzyme.

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?