• USA Home
  • M5936 - Greiner high and medium binding 384 well plates
EMAIL THIS PAGE TO A FRIEND
M5936 Sigma
Greiner high and medium binding 384 well plates
clear flat bottom, high-binding
Purchase
Properties
Related Categories 384 Well Plates, Antibody Plates, Antibody/Immunology Supplies, Cell Culture, Drug Discovery,
material   clear flat bottom
feature   high-binding
mfr. no.   Greiner 781061
Description
General description
Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?