• USA Home
  • M6167 - Macrophage Inflammatory Protein-1α from mouse

EMAIL THIS PAGE TO A FRIEND
M6167 Sigma

Macrophage Inflammatory Protein-1α from mouse

≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Synonym: MIP-1α

Purchase

Description

Analysis Note

The biological activity is measured by its ability to chemoattract cultured human monocytes or mouse BaF/3 cells transfected with human CCR5.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?