• USA Home
  • M6188 - Mimetic Yellow 2 Ligand Affinity Adsorbent

EMAIL THIS PAGE TO A FRIEND
M6188 Sigma

Mimetic Yellow 2 Ligand Affinity Adsorbent

suspension

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
form   suspension
shipped in   wet ice
storage temp.   2-8°C

Description

Application

Ligand affinity adsorbents are used in protein chromatography, affinity chromatography and dye resins. Ligand affinity adsorbents have been used to describe a simple protocol for the purification of Taq Polymerase from E. coli lysates as well as for the purification of other DNA polymerases.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 3 - Ethanol, solution
WGK Germany 
3
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?