• USA Home
  • M6255 - 3-Methylfentanyl hydrochloride, 100 μg/ml Hydrochloride

EMAIL THIS PAGE TO A FRIEND
M6255 Sigma

3-Methylfentanyl hydrochloride, 100 μg/ml Hydrochloride

Purchase

Properties

form   neat
drug control   Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?