• USA Home
  • M6286 - 4-(4-Maleimidophenyl)butyric acid N-hydroxysuccinimide ester

EMAIL THIS PAGE TO A FRIEND
M6286 Sigma

4-(4-Maleimidophenyl)butyric acid N-hydroxysuccinimide ester

Synonym: 4-(4-Maleimidophenyl)butyric acid N-succinimidyl ester, 4-(4-Maleimidophenyl)butyric acid NHS ester, N-Succinimidyl 4-(4-maleimidophenyl)butyrate, SMPB

Purchase

Description

Application

Heterobifunctional cross-linking reagent.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?