• USA Home
  • M6289 - Millipore® solid-phase extraction apparatus vacuum manifold
EMAIL THIS PAGE TO A FRIEND
M6289 Sigma
Millipore® solid-phase extraction apparatus vacuum manifold
6-place kit for 47 mm filters
Purchase
Properties
Related Categories Filtration Supplies, Labware, Vacuum Manifold More...
mfr. no.   Millipore, XX0047SPE
Description
General description
Dramatically reduces extraction times for environmental analysis. Analytes retained on a solid support can be eluted with a small quantity of solvent. Three-way valve allows sample filtration, washing, and elution without dismantling.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Standard component
Description
Product #
Add to Cart
300-ml glass funnel 6 ea SDS    
47 mm anodized aluminum spring clamp 6 ea SDS    
47 mm base with 3-way valve 6 ea SDS    
47 mm PTFE-coated stainless steel screen 6 ea SDS    
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
M4664 Nalgene® vacuum manifold, autoclavable
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?