• USA Home
  • M6312 - Greiner HLA Terasaki multiwell plates
EMAIL THIS PAGE TO A FRIEND
M6312 Sigma
Greiner HLA Terasaki multiwell plates
72 well, 1 lid per plate (space saver design)
Synonym: Greiner HLA Terasaki multiwell plates
Purchase
Properties
Related Categories Antibody Plates, Antibody/Immunology Supplies, Cell Culture Supplies, Culture Multiwell Plates, Greiner,
feature   tissue-culture treated
mfr. no.   Greiner 769190
well size   10 μL
Description
General description
HLA Microtest plates (Terasaki) from Greiner. Designed for tissue typing techniques to ensure smooth spreading of sera and cells across the flat base of the well. The surface of each well gives a microscopically precise and clear picture. All plates are stackable.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?