EMAIL THIS PAGE TO A FRIEND
M6555 Sigma

Medium 199

Purchase

Properties

Quality Level   GMP
storage temp.   2-8°C

Description

Application

Originally developed as a completely defined media formulation for the culture of primary explants. These media have broad applicability, particularly for non-transformed cells and are widely used for vaccine production, primary pancreatic explants, and lens tissues.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?