• USA Home
  • M6561 - Greiner 384 well plates, polystyrene

EMAIL THIS PAGE TO A FRIEND
M6561 Sigma

Greiner 384 well plates, polystyrene

clear polystyrene wells flat bottom, high-binding, sterile

Synonym: 384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates

Purchase

Properties

Related Categories 384 Well Plates, Antibody Plates, Antibody/Immunology Supplies, Greiner, Greiner 384-Well Multiwell Plates,
material   clear polystyrene wells flat bottom
  white polystyrene plate
sterility   non-sterile
  sterile
feature   high-binding
  lid: No
packaging   case of 40
mfr. no.   Greiner 781094
maximum volume   131 μL
size   384 wells
working volume   110 μL
color   clear
matrix active group   μClear® and tissue culture treated surface treatment

Description

General description

CELLSTAR®, 384W microplate, with an optimal working volume of 15-110 μL; maximum well capacity of 131 μL. Growth area: 10 mm(2)

Greiner multiwell plates are available in the standard multiwell plate footprint and offer excellent optical properties. These plates are DNAse /RNAse free and non-pyrogenic as deteremined by LAL method (< 0.06 EU/mL). Appropriate for general purposes, such as optical measurements, photography, and microscopy, due to the high transparency of polystyrene plates.

Other Notes

384 well flat (F) solid bottom microtiter plate, without lid, rounded square well design, alphanumeric well coding; length: 127.76 +/- 0.15 mm; width: 85.48 +/- 0.15 mm; curvature: approx.150 μm.

Suitability

White plates minimize cross talk in luminescence procedures. μClear film improves Luminescence; Fluorescence; Cell monitoring; Transmission; and Imaging by reducing background, cross-talk and autofluorescence. Suitable for cell work. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

μClear is a registered trademark of Greiner Bio-One GmbH

CELLSTAR is a registered trademark of Greiner Bio-One GmbH

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?