• USA Home
  • M6637 - 5-Methoxyindole-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
M6637 Sigma

5-Methoxyindole-2-carboxylic acid

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL6005000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?