• USA Home
  • M6643 - Myosin Calcium activated from bovine muscle

EMAIL THIS PAGE TO A FRIEND
M6643 Sigma

Myosin Calcium activated from bovine muscle

buffered aqueous glycerol solution, 0.3-1.0 units/mg protein (biuret)

Purchase

Properties

form   buffered aqueous glycerol solution
color   clear light yellow
shipped in   wet ice
storage temp.   −20°C

Description

Unit Definition

One unit will liberate 1.0 μmole of inorganic phosphorus from ATP per min at pH 9.0 at 25°C in the presence of calcium.

Physical form

solution in 50% glycerol containing 06 M KCl and 10 mM potassium phosphate buffer, pH 68

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

M1636 Myosin, Calcium activated from rabbit muscle, buffered aqueous glycerol solution, 0.5-1.5 units/mg protein (biuret)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?