• USA Home
  • M6661 - MPL® + TDM + CWS Adjuvant

EMAIL THIS PAGE TO A FRIEND
M6661 Sigma

MPL® + TDM + CWS Adjuvant

oil

Synonym: Monophosphoryl-lipid A + Trehalose dicorynomycolate + Cell wall skeleton adjuvant

Purchase

Properties

Related Categories Adjuvant Reagents, Antibodies, Antibody Purification and Characterization, Supplementary Products
form   oil
storage temp.   2-8°C

Description

General description

Each vial contains 0.5 mg monophosphoryl lipid A from S. minnesota (MPL), 0.5 mg trehalose dicorynomycolate (TDM) and 0.5 mg cell wall skeleton from a tubercule bacillus (CWS) in 2% oil-Tween 80-water. The CWS provides additional immunopotentiating activity.

Application

Ribi Adjuvant System (RAS) for use in rabbits, goats and larger animals.
RAS is a stable oil-in-water emulsion that may be used as an alternative to the water-in-oil emulsions. These adjuvants are derived from bacterial and mycobacterial cell wall components that have been prepared to reduce the undesirable side effects of toxicity and allergenicity, but still provide potent stimulus to the immune system.

Legal Information

MPL is a registered trademark of Corixa Corporation

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

T2684 TiterMax® Gold Adjuvant, liquid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?