• USA Home
  • M6812 - Greiner 96 well 1 x 8 strip plates

EMAIL THIS PAGE TO A FRIEND
M6812 Sigma

Greiner 96 well 1 x 8 strip plates

high binding, clear polystyrene wells flat bottom (1×12 well strips)

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Purchase

Properties

Related Categories 96 Well Plates, Antibody Plates, Antibody/Immunology Supplies, Greiner, Greiner 96-Well Multiwell Plates,
material   polystyrene
  clear polystyrene wells flat bottom (1×12 well strips)
sterility   non-sterile
packaging   case of 100
mfr. no.   Greiner 701071
maximum volume   388 μL
size   96 wells
working volume   350 μL
color   clear plate strips
matrix active group   high-binding surface treatment

Description

General description

MICROLON® 600, 96W 8xF12 Strip Microplate, working volume 20-350 μL; total well volume 388 μL.

Other Notes

96 well flat (F) solid bottom microplate, high-binding, F12 strips mounted in frame, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm.

Suitability

UsefμL in immunological assays. MICROLON 600 is a hydrophobic medium binding surface that will bind up to 600 ng/cm2 of non-polar proteins and peptides. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?