• USA Home
  • M6813 - Mimetic Blue SA Ligand Affinity Adsorbent

EMAIL THIS PAGE TO A FRIEND
M6813 Sigma

Mimetic Blue SA Ligand Affinity Adsorbent

suspension

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
form   suspension
shipped in   wet ice
storage temp.   2-8°C

Description

Application

Ligand affinity adsorbents are used in protein chromatography, affinity chromatography and dye resins. Ligand affinity adsorbents have been used to describe a simple protocol for the purification of Taq Pol from E. coli lysates as well as for the purification of other DNA polymerases.

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 3 - Ethanol, solution
WGK Germany 
1
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?