• USA Home
  • M7032 - Malic Dehydrogenase from Thermus flavus

EMAIL THIS PAGE TO A FRIEND
M7032 Sigma

Malic Dehydrogenase from Thermus flavus

lyophilized powder, ≥50 units/mg solid

Synonym: L-Malate: NAD+ oxidoreductase, MDH

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of oxalacetate and β-NADH to L-malate and β-NAD per min at pH 7.5 at 25 °C.

Physical form

Contains dextran

Application

Malic dehydrogenase has been used in a study to assess electron transport chain activity in mitochondria from human skeletal muscle. It has also been used in a study to investigate activities of enzymes and ammonia in serum of rats with fluoride hyperglycemia.

General description

Malic dehydrogenase from Thermus flavus is stable and active at 90 ° C.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?