• USA Home
  • M7035 - Components Of N-Linked Oligosaccharide Sequencing Kit

EMAIL THIS PAGE TO A FRIEND
M7035 Sigma

Components Of N-Linked Oligosaccharide Sequencing Kit

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?