• USA Home
  • M7146 - DL-Mandelic acid hemimagnesium salt

EMAIL THIS PAGE TO A FRIEND
M7146 Sigma

DL-Mandelic acid hemimagnesium salt

~95%

Synonym: ±-α-Hydroxyphenylacetic acid

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
InChI Key   RPONRISSUINNRF-UHFFFAOYSA-L
assay   ~95%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP

From our library of Articles, Sigma-Aldrich presents GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?