• USA Home
  • M7156 - Methyl 13-methylmyristate

EMAIL THIS PAGE TO A FRIEND
M7156 Sigma

Methyl 13-methylmyristate

≥98% (capillary GC)

Synonym: 13-Methyltetradecanoic acid methyl ester

  • CAS Number 5129-59-9

  • Empirical Formula (Hill Notation) C16H32O2

  • Molecular Weight 256.42

  •  MDL number MFCD00083432

  •  PubChem Substance ID 24897144

Purchase

Properties

Related Categories Biochemicals and Reagents, Branched fatty acids, Fatty Acyls, Fatty Esters, Lipids,
InChI Key   OGGUSDOXMVVCIX-UHFFFAOYSA-N
assay   ≥98% (capillary GC)
storage temp.   2-8°C

Description

Packaging

10 mg in ampule

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?