• USA Home
  • M7184 - Matrix Metalloproteinases and TIMPs
EMAIL THIS PAGE TO A FRIEND
M7184 Sigma
Matrix Metalloproteinases and TIMPs
  •  ISBN-10 0-19-850268-0
  •  ISBN-13 978-0-19-850268-5
Purchase
Properties
Related Categories Application Index, Biochemicals and Reagents, Biochemistry Books, Books and Software, Cell Signaling Enzymes,
publication info   J.F. Woessner and H. Nagase, ed., Oxford University Press, 2000, 240 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This book contains comprehensive data about matrix metalloproteinases (MMPs), enzymes which break down the extracellular matrix. It covers their sequences, three-dimensional structure, activation, substrates, specificity, inhibition, biological roles and function, along with protein sequence alignments. MMPs are regulated by their inhibitors (TIMPs). This volume contains sequences, three-dimensional structures and biological roles of TIMPs along with over 1700 references on MMPs and TIMPs.
Table of Contents
Abbreviations; Introduction; MMP sequences; TIMP sequences; Three dimensional structures of the MMPs and TIMPs; Activation of the zymogen forms of MMPs; Protein substrates of the MMPs; Specificity requirements of the MMPs; Inhibition of the MMPs; Biological roles of the MMPs; Biological roles of the TIMPs; Bibliography; Protein sequence alignments
Price and Availability
Biomedical Applications
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?